Tisk…
Industrial gloves, Ultrane 550
Dodavatel: MAPA
34550540 34550546 34550547 34550548 34550549
112-5628EA 1020 CZK
112-5628 112-5624 112-5625 112-5626 112-5627
Industrial gloves, Ultrane 550
Rukavice
These polyurethane foam gloves with seamless textile support and ventilated back provide light protection.
  • Excellent abrasion resistance: longer lasting
  • Excellent tactile sensitivity, due to reduced glove thickness
  • Absorption of perspiration ensured by the cellular structure of the polyurethane
  • Seamless knit lining for additional comfort
Suitable for precision work, such as intricate assembly, sorting small parts, fitting small screws and fasteners, electronic component assembly (displays, LEDs, etc.), high-frequency antennae riveting, microprocessor handling and cosmetics preparation.
Certifikace: EN 388:2016 (4131X); Cat. II