Tisk…
Neoprene gloves, Microflex® NeoTouch® 25-101 / 25-201
Dodavatel: Ansell
25-101/6.5-7 25-101/7.5-8 25-101/8.5-9 25-101/9.5-10 25-201/6.5-7 25-201/7.5-8 25-201/9.5-10 25-201/8.5-9
112-0027EA 1020 CZK
112-0027 112-0028 112-0029 112-0030 112-0031 112-0032 112-0034 112-0033
Neoprene gloves, Microflex® NeoTouch® 25-101 / 25-201
Rukavice
These neoprene single-use gloves with beaded cuffs provide excellent resistance to acids and bases.
  • Prevents Type I allergies and skin reactions, free of plasticisers
  • Excellent comfort, similar to that of latex
  • Polyurethane inner coating ensures easy donning
  • Textured fingertips provide a secure grip in both dry and wet conditions
  • Green colour for easy identification
Ideal for use in the pharmaceutical and chemical industries.
Length: 240 or 290 mm
Thickness: 0,13 mm
Colour: Green
EN 374, Cat. III; AQL 1.5
Certifikace: EN ISO 374-1:2016More Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Obouruké/jednoruké Obouruké
Jednorázové/znovupoužitelné Jednorázové
Latex/bez latexu Latex-free
Pudrované/nepudrované Nepudrované
Provedení Textured fingers
Typ manžety Rolovaná
Norma EN EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
Antistatický +