Tisk…
Dropping funnels, cylindrical, with pressure balance
Dodavatel: Lenz Laborglas GmbH & CO.KG
04354914 04355814 04357014 04353714 04352814 04352211 04352811 04353711
271-5164EA 3090 CZK
271-5164 271-5165 271-5166 271-5163 271-5162 271-5154 271-5152 271-5153
Dropping funnels, cylindrical, with pressure balance
Nálevky Nálevky přikapávací
DURAN®, borosilicate glass, clear, with glass key.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint cone and socket, with divisions for approximate volume reading. Stopcock with thread guard.