Tisk…
Funnels with short stem
Dodavatel: KARTELL
172 173 174 175 176 177 178 4171 4179
221-1521EA 114 CZK
221-1521 221-1522 221-1523 221-1524 221-1525 221-0080 221-0081 221-1263 221-0082
Funnels with short stem
Nálevky Nálevky filtrační Nálevky k filtraci
HDPE, translucent.
  • Good chemical resistance
  • With raised lip to prevent overflowing
  • External ribs for unobstructed removal of air when filtering
With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.