Tisk…
Funnels, analytical
Dodavatel: KARTELL
145 146 147 148 150 152 153 155 156
221-1811EA 79.7 CZK
221-1811 221-1812 221-1813 221-1814 221-1815 221-1816 221-1520 221-1818 221-1819
Funnels, analytical
Nálevky Nálevky filtrační Nálevky analytické
PP, translucent.
  • Inner ribs at an angle of 58° to accelerate the filtering process
  • Outer ribs prevent air from being trapped under the filter paper
  • Autoclavable
With inner and outer ribs. Cone angle 60°.