Tisk…
Forceps
Dodavatel: KARTELL
500 501 503
232-2000EA 978 CZK
232-2000 232-2001 232-2003
Forceps
Pinzety
Polyoxymethylene (POM), yellow. Multipurpose forceps for a wide range of applications.
  • Good chemical resistance, not suitable for use with strong acids and solvents
  • Autoclavable
  • Robust, pliable, self-sprung