Tisk…
Flow indicators
Dodavatel: BURKLE
8720-2002 8720-2003 8720-2005 8720-1003 8720-1002 8720-1005
618-1121EA 423 CZK
618-1121 618-1122 618-1120 618-1112 618-1111 618-1110
Flow indicators
Indikátory průtoku
These paddle wheel flow indicators give a clear visualisation of the flow rates of liquids or gas in tubing.
  • Colour coding - Red/Black/Blue
  • Flow indicator in either direction of flow and can be mounted at any angle
  • Paddle wheel impeller starts to rotate at about 150 ml/minute
LiquiMobil in SAN: Suitable for coolant circuits and contributes to reduction of water consumption appreciably. Maximum operating temperature 30 °C.
CheMobil in PMP: Chemical resistance makes for use with a great number of fluids. CheMobil is steam sterilisable. Maximum operating temperature 60 °C.
Acceptable maximum pressure 2 bar
Connector o.d.: 6 - 11 mm
Dimensions: 88×15×40 mm
Upozornení: LiquiMobil is not for use with aggressive liquids - CheMobil is not appropriate for strong acids, bases and solvents.