Tisk…
Flasks, round bottom with three necks and standard ground joints
Dodavatel: Lenz Laborglas GmbH & CO.KG
03303249 03303258 03303270 03304137 03304149 03304158 03304170 03304173 03304237 03304249 03304258 03304270 03304437 03304449 03304458 03304470 03304473 03304479 03304482 03305479 03305482 03304476 03305470 03305473 03305485 03305488 03304485 03304176 03305476 03303237 03303273 03304179 03304185 03304273 03304488 03305494
201-1184EA 1790 CZK
201-1184 201-1185 201-1186 201-1521 201-1522 201-1524 271-1526 271-1529 LENZ03304237 LENZ03304249 201-1188 LENZ03304270 LENZ03304437 201-1528 201-1525 201-1527 201-1530 201-1529 201-0335 271-1532 271-1533 201-1046 271-1528 271-1531 271-1534 271-1535 201-1047 LENZ03304176 LENZ03305476 201-0523 201-1187 LENZ03304179 LENZ03304185 201-1189 201-1048 LENZ03305494
Flasks, round bottom with three necks and standard ground joints
Baňky Baňky s kulatým dnem
DURAN®, borosilicate glass, with two parallel side necks, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Central neck and two parallel side necks. Matt white labelling area.