Tisk…
Fernbach culture flasks, DURAN®
Dodavatel: DWK Life Sciences
215014307 215116203
391-0330EA 1130 CZK
391-0330 391-0320
Fernbach culture flasks, DURAN®
Baňky Baňky kultivační
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With beaded rim. Providing the large liquid surface area necessary for growing microorganisms in liquid culture media. Provides for faster growth through improved ventilation.