Tisk…
Erlenmeyer flasks, narrow neck, with flared rim, Nalgene®
Dodavatel: Thermo Fisher Scientific
4102-0125 4102-1000 4102-2000 4103-0125 4103-0250 4102-0250
214-7102EA 4290 CZK
214-7102 214-7105 214-7106 214-7122 214-7123 214-7103
Erlenmeyer flasks, narrow neck, with flared rim, Nalgene®
Baňky Baňky dle Erlenmeyera
PPCO, translucent or PC, transparent.
  • Much safer than glass flasks when used with shakers
  • Autoclavable. Repeated autoclaving of PC flasks can eventually impair their mechanical strength
  • Smooth surfaces for fast, easy cleaning
Flasks made from PPCO with permanent graduations suitable for general laboratory use. The level of the liquid content is easy to read. Flasks made from PC are designed especially to be used in creating and storing nutrient media, and for many culturing procedures. The liquid level can be read easily and accurately.