Tisk…
Desiccator lids, DURAN®
Dodavatel: DWK Life Sciences
244104607 244105706 244106102 244106608 244106908 244204605 244205704 244206109 244206606 244206906 244405709 244406105 244406602 244406902
467-1160EA 2670 CZK
467-1160 467-1175 467-1180 467-1185 467-1190 467-1260 467-1275 467-1280 467-1285 467-1290 467-1375 467-1380 467-1385 467-1390
Desiccator lids, DURAN®
Víčka Víka k exsikátorům
DURAN®, borosilicate glass 3.3, clear plain lids for desiccators. Lids have either knob or NS-connection for stop cork (Novus) or thread connection for stop cork (Mobilex).
Certifikace: According to DIN ISO 13130