Tisk…
Crucibles, quartz
Dodavatel: NEUBERT VOLUME GLASSWAERE
8500-006 8505-050 8505-075 8505-120
459-0160EA 1570 CZK
459-0160 459-0161 459-0162 459-0163
Crucibles, quartz
Kelímky
Transparent quartz glass.
  • Withstand constant maximum temperature of 1100 °C, short term use at 1300 °C
Lids are available separately.