Tisk…
HPLC columns, ZirChrom®
Dodavatel: HICHROM
EZ01-1546 ZR03-1546
554-7686EA 44010 CZK
554-7686 554-7684
HPLC columns, ZirChrom®
Kolony chromatografické
Zirconia bonded phases offer unique selectivity relative to conventional silica based columns, combined with extreme chemical and thermal stability. Unlike polymeric phases, zirconia will not shrink or swell as a function of eluent organic content or ionic strength.
  • Porous zirconia based reverse phase materials
  • High pH and temperature stability
  • Fast solvent equilibration