Tisk…
HPLC columns, LiChrosorb®
Dodavatel: Merck
1.50318.0001 1.50332.0001 1.50333.0001 1.50334.0001 1.50388.0001 1.50432.0001 1.50433.0001 1.51343.0001 1.51345.0001 1.51349.0001 1.51351.0001 1.51353.0001 1.51354.0001 1.51355.0001 1.51356.0001
1.50318.0001EA 25230 CZK
1.50318.0001 1.50332.0001 1.50333.0001 1.50334.0001 1.50388.0001 1.50432.0001 1.50433.0001 1.51343.0001 1.51345.0001 1.51349.0001 1.51351.0001 1.51353.0001 1.51354.0001 1.51355.0001 1.51356.0001
HPLC columns, LiChrosorb®
Kolony chromatografické
LiChrosorb® is one of the most successful HPLC packing materials. It is a totally porous, irregular particle silica gel produced with pore sizes of 60 and 100 Å. LiChrosorb® HPLC columns are available either as a cartridge, requiring re-usable endfittings, or as a complete end-fitted column.
If guard columns are required, use LiChrospher® guard cartridges listed in LiChrospher® section.