Tisk…
Overalls, Monotex®, PP
Dodavatel: Zellstoff-Vertriebs-GmbH & Co. KG
30733-00 30734-00 30735-00
114-3425EA 148 CZK
114-3425 114-3426 114-3427
Overalls, Monotex®, PP
Jednorázové oděvy
The overall is made from 100% PP and ensures protection against coarse, non aggressive, and non toxic dirt or dust particles. It features a hood with elastic band, zip with protective flap, elasticated wrists and ankles.
  • Comfortable and breathable material
  • Ideal overall for visitors
  • Should not be used in hazardous areas
Colour: White
Certifikace: Cat. I
Balení: Individually packed.