Tisk…
Laboratory coats, secutex® pro, PP/PE
Dodavatel: Zellstoff-Vertriebs-GmbH & Co. KG
30203-90 30204-90 30205-90
114-3433EA 319 CZK
114-3433 114-3434 114-3435
Laboratory coats, secutex® pro, PP/PE
Jednorázové oděvy
Laboratory coats made from laminated PP/PE feature doubled collar, press-stud fasteners and two side pockets.
  • Anti-static
  • Solid material allows water vapour to escape for increased comfort
  • Protection against dust and splashes of aggressive liquids
Colour: white