Tisk…
Screw caps for square bottles, DURAN®
Dodavatel: DWK Life Sciences
293031901 293032803 293033208 293033508 293011908 293012801 293021909
217-9204EA 325 CZK
217-9204 217-9205 217-9206 217-9207 217-9200 217-9201 217-9202
Screw caps for square bottles, DURAN®
Uzávěry Uzávěry k láhvím
PP, red or blue.
  • Standard caps, blue, with snap ring for narrow mouth and wide mouth bottles
  • Safety caps with valve (valve cap opens with 0,1 bar overpressure), for narrow mouth bottles
  • Very good chemical and temperature resistance, leakproof
Tamper evident caps for narrow mouth and wide mouth square bottles with DIN thread.