Tisk…
Screw caps, for premium laboratory bottles, DURAN®
Dodavatel: DWK Life Sciences
1088678 1088679 1129600
215-0090EA 656 CZK
215-0090 215-0091 215-2856
Screw caps, for premium laboratory bottles, DURAN®
Uzávěry Uzávěry k láhvím
TPCh260, colourless.
  • Highly chemically resistant material (TPCh260), no added colours (any interference with the medium is avoided), temperature resistant from −196 to +200 °C
  • PTFE coated silicone seal
  • Drip-free and safe due to pouring ring, which can be ordered separately
The Premium screw caps meet the highest demands for storing and packaging products from the pharmaceutical, chemical and food industries. Ideal for dry sterilisation at high temperatures. Suitable for all GL 25/GL 45 threads.
Complies with USP/EP/FDA standards.
With printedMore Product Information