Tisk…
Burette clamps
Dodavatel: USBECK Laborgeräte
2841 2842
241-7451EA 567 CZK
241-7451 241-7452
Burette clamps
Svorky Svorky pro běžné účely
Zinc die-cast, nickel plated. For simple, secure fixing of burettes.
  • Bosshead for fixing to stand rods
  • Jaws spanning distance from 0 to 40 mm
  • Clamping jaws with plastic coating to protect burettes