Tisk…
Kartony ze spalovny biohazardu, SP Bel-Art
Dodavatel: Bel-Art Products, a Part of SP
F132050001 F132050002
119-0200EA 5840 CZK
119-0200 119-0201
Kartony ze spalovny biohazardu, SP Bel-Art
Krabice Krabice odpadkové
Corrugated cardboard.
  • Snug fitting lid has flap through which waste is inserted
  • When carton is filled, internal safety cap is pulled into place and the entire unit is ready for incineration
Carton with integral biohazard bag in HDPE.
Informace o dodávce: Delivered unassembled. Assembly takes a few seconds.