Tisk…
Kryoboxy, Nalgene®
Dodavatel: Thermo Fisher Scientific
5055-5002 5055-5005 5055-5015
525-0050EA 1481 CZK
525-0050 525-0051 479-0012
Kryoboxy, Nalgene®
Krabice Kryoboxy
PC with foam insert.
  • Accepts writing with cryo-markers
  • Autoclavable
For temperature storage of cryovials from –150 to +121 °C (not in liquid phase of liquid nitrogen).
Transparent lid with printed grid permits viewing of contents and are keyed to prevent misalignment.