Tisk…
Láhve, úzké hrdlo, kulaté, PFA, se šroubovacím uzávěrem
Dodavatel: VITLAB
109297 109397 108297 108397 108497
215-4591EA 2310 CZK
215-4591 215-4592 215-4596 215-4597 215-4598
Láhve, úzké hrdlo, kulaté, PFA, se šroubovacím uzávěrem
Láhve Láhve plastové
PFA, translucent, with PFA screw cap.
  • Maximum chemical resistance
  • Heat resistant from −200 to +250 °C, autoclavable
  • Leakproof
Bottles with high resistance to extreme temperatures, chemicals and corrosion. Inert to practically all chemicals with the exception of molten alkaline metals, fluorine compounds and complex halogen compounds at high temperatures and under pressure. Ideal for high purity liquids.