Tisk…
Bottles, wide neck, with screw cap, Mason type, Nalgene®
Dodavatel: Thermo Fisher Scientific
2115-0500 2115-1000 2115-2000 2115-3000
215-7851EA 2920 CZK
215-7851 215-7852 215-7853 215-7854
Bottles, wide neck, with screw cap, Mason type, Nalgene®
Láhve Láhve plastové
PPCO, translucent with PP screw cap.
  • Very good chemical resistance
  • Heat resistant up to 121 °C, autoclavable
  • Leakproof with ring for tamper evident heat shrink bands
Bottles have 70 mm necks that accept fixtures or closures with Mason jar threads. Also fit on mixers equipped with the special Mason thread.