Tisk…
Bottles, wide neck, with screw cap, Nalgene®
Dodavatel: Thermo Fisher Scientific
2103-0001 2103-0002 2103-0004 2103-0008 2103-0016 2103-0032
215-7211EA 491 CZK
215-7211 215-7212 215-7213 215-7214 215-7215 215-7216
Bottles, wide neck, with screw cap, Nalgene®
Láhve Láhve plastové
LDPE, translucent with PP screw cap.
  • Good chemical resistance, heat resistant up to +80 °C
  • Leakproof closure, so they are suitable for use with liquids
  • Flexible, excellent impact resistance
Can be used anywhere in the laboratory and for collecting samples. Wide opening allows rapid filling and dispensing of powders, pastes and liquids.