Tisk…
Bottles, narrow neck, round, PP, with standard ground stoppers
Dodavatel: VITLAB
100594 100694
215-6844EA 1740 CZK
215-6844 215-6845
Bottles, narrow neck, round, PP, with standard ground stoppers
Láhve Láhve plastové
PP, translucent, with HDPE stoppers.
  • Very good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
Perfect for sample collection, storage and transport. Ideal for liquids. Can be filled with hot substances.