Tisk…
Bottles, narrow neck, round, PE, with thread
Dodavatel: KARTELL
15801 15821 15841 15861 15881 15901
215-1329EA 95 CZK
215-1329 215-1330 215-1331 215-1332 215-1333 KART15901
Bottles, narrow neck, round, PE, with thread
Láhve Láhve plastové
Polyethylene translucent.
  • Ideal for water and oils, no shoulders to restrict flow
  • Graduated so they can be used as rough measures
Bottles can be secured using shoulder and cap tag to avoid sample contamination in transit.
Certifikace: DlN 13316 and 168
Upozornení: Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904.