Tisk…
Dilution bottles, Square, PSF/PPCO, with screw caps, Nalgene®
Dodavatel: Thermo Fisher Scientific
2500-0280 2500-0380 2505-0380
215-7801EA 3560 CZK
215-7801 215-7802 215-7812
Dilution bottles, Square, PSF/PPCO, with screw caps, Nalgene®
Láhve Láhve plastové
Polysulphone PSF, crystal clear or polypropylene copolymer PPCO, translucent, with PP screw cap.
  • PSF: Outstanding resistance to weak acids, bases and all aqueous solutions/PPCO: Outstanding resistance to strong acids, alcohols and bases
  • PSF: Temperature resistant from −100 to +165 °C/PPCO: Temperature resistant from −40 to +121 °C
  • PSF: Autoclavable, use with Tween shortens service life/PPCO: Autoclavable, sustains repeated cleaning and autoclaving with Tween
Compliant with the dilution methods described in the standard methods for waterMore Product Information