Tisk…
Demižóny, široký krk, PC, Clearboy™, Nalgene®
Dodavatel: Thermo Fisher Scientific
DS2213-0020 DS2213-0050
216-5101EA 6920 CZK
216-5101 216-5102
Demižóny, široký krk, PC, Clearboy™, Nalgene®
Láhve Demižóny
PC, transparent with white PP screw cap.
  • Good chemical resistance
  • Thermal resistance from −135 to +135 °C, autoclavable
  • Light, extremely impact resistant, leakproof
Ideal for storing solids and for use at extreme temperatures. Wide opening (90 mm diameter) for easy filling and decanting or cleaning. Calibrated in gallons and litres. Sturdy stainless steel handle.