Tisk…
Shazovací lahve, LDPE, Nalgene®
Dodavatel: Thermo Fisher Scientific
2750-9025 2750-9050 2750-9125 2751-9025 2751-9050 2751-9125
215-7171EA 1670 CZK
215-7171 215-7172 215-7173 215-1058 215-1059 215-1060
Shazovací lahve, LDPE, Nalgene®
Láhve Lahvičky kapací
LDPE, translucent or LDPE, white with white PP screw cap and LDPE dosing tip.
  • Good chemical resistance, heat resistant up to +80 °C
  • Drop-control tip snaps securely into place, drops are dispensed individually
  • Flexible for easy handling, leakproof
  • Delivers 40 µl drops (based on water; viscosity affects drop size)
These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic andMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable Low Manufacturing Impact
Certifikát ochrany životního prostředí NAR Certificate of Retirement