Tisk…
Bottles, wide neck, HDPE, with screw cap
Dodavatel: KARTELL
1640 1642
216-2301EA 898 CZK
216-2301 216-2302
Bottles, wide neck, HDPE, with screw cap
Láhve Láhve plastové
HDPE, translucent with blue screw cap.
  • Good chemical resistance
  • Heat resistant up to +80 °C (+120 °C for short periods)
  • Heavy walled, very tough, insert plug and deeper closure
Ideal for storing powders, pastes, and liquids. Easy filling and dispensing. Includes carrying handle. Suitable for food contact as compliant with FDA standard.