Tisk…
Beakers, tall form, DURAN®
Dodavatel: DWK Life Sciences
211161704 211162409 211162906 211163602 211164101 211164804 211165303 211165406 211166308 211166805
213-1154EA 292 CZK
213-1154 213-1155 213-1156 213-1157 213-1158 213-1159 213-1160 213-1161 213-1162 213-1163
Beakers, tall form, DURAN®
Kádinky
DURAN®, borosilicate glass 3.3, clear, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area.
With printed retrace code (batch identification).
ISO 3819.