Tisk…

Sodium diphenylamine-4-sulphonate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium Diphenylaminesulphonate, Sodium N-phenylsulphanilate, Sodium (4-anilinobenzenesulphonate), Diphenylamine-4-sulphonic acid, sodium salt

M-Clarity™ quality level = MQ200

242969-25G 242969-5G
SIAL242969-25GEA 10580 CZK
SIAL242969-25G SIAL242969-5G
Sodium diphenylamine-4-sulphonate, Sigma-Aldrich®
Sodium diphenylamine-4-sulphonate
Vzorec: C₁₂H₁₀NNaO₃S
MW: 271,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036468
CAS číslo: 6152-67-6
EINECS: 228-165-6
Merck Index: 13,07397

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR