Murexide, Supelco®
Dodavatel: Merck
Synonyms:
Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
M-Clarity™ quality level = MQ200
1.06161.0005
1.06161.0025
1.06161.0005EA
2150
CZK
1.06161.0005
1.06161.0025
Murexide, Supelco®
Murexide
Vzorec:
C₈H₈N₆O₆ MW: 284,19 g/mol Bod tání: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012777 CAS číslo: 3051-09-0 EINECS: 221-266-6 Merck Index: 13,06330 |
Výsledky specifikačního testu
Solubility | 1 g/l (20 °C) |
Molar Mass | 284.19 g/mol |
Bulk Density | 330 kg/m3 |
pH | 5 (1 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...