Tisk…

Methylthymol blue sodium salt

Dodavatel: Merck

Synonyms: Sodium 2,2',2'',2'''-((((1,1-dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetate, Methylthymol Blue tetrasodium salt

M-Clarity™ quality level = MQ200

1.06084.0001 1.06084.0005
1.06084.0001EA 2660 CZK
1.06084.0001 1.06084.0005
Methylthymol blue sodium salt
Methylthymol blue sodium salt
Vzorec: C₃₇H₄₀N₂Na₄O₁₃S
Storage Temperature: Ambient
MDL Number: MFCD00148905
CAS číslo: 1945-77-3
EINECS: 217-743-3

Objednat

Výsledky specifikačního testu

Solubility 460 g/l (20 °C)
Melting Point 844.76 g/mol
Molar Mass 570 kg/m3
pH 6.2 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR