2-Methyl-1-butene-3-yne ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Methylbut-1-en-3-yne
L06111.06
L06111.14
L06111.06EA
2860
CZK
L06111.06
L06111.14
2-Methyl-1-butene-3-yne ≥97%
2-Methyl-1-butene-3-yne
Purity: 97%
CAS No.: 78-80-8
Molecular Formula: C5H6
Formula Weight: 66.10
Storage Temperature: 2°C to 8°C
Flash Point: -45°(-49°F)
Refractive Index: 1.4180
Odour: Pungent
Melting Point: -113°
Boiling Point: 32-33°
Beilstein Reference: 969175
UN No.: 3295
MDL No.: MFCD00008599
Vzorec:
C₅H₆ MW: 66,1 g/mol Bod varu: 32…33 °C Bod tání: –113 °C Hustota: 0,694 Bod vzplanutí: –45 °C (–49 °F) |
MDL Number:
MFCD00008599 CAS číslo: 78-80-8 EINECS: 201-144-9 UN: 3295 ADR: 3,I |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...