Tisk…

1H-1,2,4-Triazole-3-thiol ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 3-Mercapto-1,2,4-triazole

A13120.06 A13120.14 A13120.22
A13120.06EA 1090 CZK
A13120.06 A13120.14 A13120.22
1H-1,2,4-Triazole-3-thiol ≥98%
1H-1,2,4-Triazole-3-thiol

Purity: 98% CAS No.: 3179-31-5 Molecular Formula: C2H3N3S Formula Weight: 101.13 Melting Point: 217-224° Beilstein Reference: 107731

Vzorec: C₂H₃N₃S
Bod tání: 217…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00005229
CAS číslo: 3179-31-5
EINECS: 221-656-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR