Tisk…

Copper(II) hydroxide stabilised, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cupric hydroxide, Cuprichydroxide, Copper dihydroxide

M-Clarity™ quality level = MQ100

289787-1KG 289787-250G
SIAL289787-1KGEA 4520 CZK
SIAL289787-1KG SIAL289787-250G
Copper(II) hydroxide stabilised, Sigma-Aldrich®
Copper(II) hydroxide
Vzorec: H₂CuO₂
MW: 97.56
Hustota: 3.37 g/cm³ (20 °C)
MDL Number: MFCD00010968
CAS číslo: 20427-59-2
EINECS: 243-815-9
UN: 3288
ADR: 6.1 III
Merck Index: 12,02709

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR