Tisk…

Carboxymethyl cellulose sodium salt, high viscosity, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

M-Clarity™ quality level = MQ200

C5013-1KG C5013-2.5KG C5013-500G
SIALC5013-1KGEA 9050 CZK
SIALC5013-1KG SIALC5013-2.5KG SIALC5013-500G
Carboxymethyl cellulose sodium salt, high viscosity, Sigma-Aldrich®
Carboxymethyl cellulose sodium salt
Vzorec: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS číslo: 9004-32-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR