Tisk…

4-Aminophthalic acid 98%

Dodavatel: Thermo Scientific
Varování

L03333.03 L03333.06
L03333.03EA 2020 CZK
L03333.03 L03333.06
4-Aminophthalic acid 98%
4-Aminophthalic acid

Purity: 98% CAS No.: 5434-21-9 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Odour: Odourless Melting Point: 300° Beilstein Reference: 2723405 MDL No.: MFCD00013985

Vzorec: C₈H₇NO₄
MW: 181,15 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013985
CAS číslo: 5434-21-9
EINECS: 226-596-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR