Xylenol orange tetrasodium salt, Supelco®
Dodavatel: Merck
Synonyms:
Xylenol orange water soluble, 2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid, sodium salt (1:4)
M-Clarity™ quality level = MQ200
1.08677.0001
1.08677.0005
1.08677.0001EA
3850
CZK
1.08677.0001
1.08677.0005
Xylenol orange tetrasodium salt, Supelco®
Xylenol orange tetrasodium salt
Vzorec:
C₃₁H₂₈N₂Na₄O₁₃S MW: 760.59 Bod tání: ∼210 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044293 CAS číslo: 3618-43-7 EINECS: 222-805-8 |
Výsledky specifikačního testu
Solubility | 510 g/l (20 °C) |
Molar Mass | 760.59 g/mol |
Bulk Density | 250 kg/m3 |
pH | 8.1 (10 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...