Methyl cellulose, 8000 cPs (C=2%, H₂O)
Dodavatel: Thermo Fisher Scientific
Synonyms:
Cellulose methyl ether
43483.22
43483.36
43483.22EA
1930
CZK
43483.22
43483.36
Methyl cellulose, 8000 cPs (C=2%, H₂O)
Methyl cellulose
CAS No.: 9004-67-5
Physical Form: Powder
Odour: Odourless
Merck Reference: 14,6040
MDL No.: MFCD00081763
Storage Temperature:
Ambient |
MDL Number:
MFCD00081763 CAS číslo: 9004-67-5 Merck Index: 14,6040 |