Tisk…

Methyl cellulose, 400 cP (C=2%, H₂O) USP, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cellulose methyl ether

M-Clarity™ quality level = MQ400

M0430-100G M0430-500G
SIALM0430-100GEA 7220 CZK
SIALM0430-100G SIALM0430-500G
Methyl cellulose, 400 cP (C=2%, H₂O) USP, Sigma-Aldrich®
Methyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS číslo: 9004-67-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR