Tisk…
Methyl cellulose, 400 cP (C=2%, H₂O), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Cellulose methyl ether
M-Clarity™ quality level = MQ200
M0262-100G M0262-1KG M0262-250G M0262-500G
SIALM0262-100GEA 4060 CZK
SIALM0262-100G SIALM0262-1KG SIALM0262-250G SIALM0262-500G
Methyl cellulose, 400 cP (C=2%, H₂O), Sigma-Aldrich®
Methyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS číslo: 9004-67-5