Methyl cellulose, 4000 mPa.S (2% H₂O), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Cellulose methyl ether
M-Clarity™ quality level = MQ200
M0512-1KG
M0512-100G
M0512-250G
M0512-500G
SIALM0512-1KGEA
13430
CZK
SIALM0512-1KG
SIALM0512-100G
SIALM0512-250G
SIALM0512-500G
Methyl cellulose, 4000 mPa.S (2% H₂O), Sigma-Aldrich®
Methyl cellulose
1 mPa.s = 1cP
Storage Temperature:
Ambient |
MDL Number:
MFCD00081763 CAS číslo: 9004-67-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...