Tisk…
Methyl cellulose, 25 mPa.S (2% H₂O), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
M6385-100G M6385-250G M6385-500G
SIALM6385-100GEA 3960 CZK
SIALM6385-100G SIALM6385-250G SIALM6385-500G
Methyl cellulose, 25 mPa.S (2% H₂O), Sigma-Aldrich®
Methyl cellulose
1 mPa.s = 1cP
Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS číslo: 9004-67-5