Tisk…
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ400
E4378-25G E4378-100G E4378-10G E4378-1KG E4378-250G E4378-500G
SIALE4378-25GEA 6840 CZK
SIALE4378-25G SIALE4378-100G SIALE4378-10G SIALE4378-1KG SIALE4378-250G SIALE4378-500G
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid), Sigma-Aldrich®
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)
Vzorec: C₁₄H₂₄N₂O₁₀
Bod tání: 240…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00004291
CAS číslo: 67-42-5
EINECS: 200-651-2
Merck Index: 13,03560