Tisk…
Disposable Protective Gloves, U-Fit
Dodavatel: UVEX
6016707 6016708 6016709 6016710
UVEX6016707EA 405 CZK
UVEX6016707 UVEX6016708 UVEX6016709 UVEX6016710
Disposable Protective Gloves, U-Fit
Rukavice
The uvex u-fit disposable protective glove is particularly impressive due to its good mechanical resistance. It is flexible and robust, provides good protection against dirt and oil - and is therefore perfect for light precision assembly work.
  • Thin and reliable NBR disposable glove (0,10 mm)
  • NBR (nitrile rubber)
  • Excellent tactile and fingertip sensitivity
  • Very good fit
  • Very high flexibility
Good grip due to the roughened glove surface and very good mechanical strength. Reliable splash protection when handling chemicals such as acids, alkalis, solids or aqueous salt solutions.
Silicone-free according to impression test. Suitable for handling diluted pesticides.
Certifikace: EN ISO 374-1:2016/Type B (KPT)