Tisk…

1-Iodoperfluoropropane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Perfluoropropyl iodide, Heptafluoro-1-iodopropane, 1,1,2,2,3,3,3-Heptafluoro-1-iodopropane

M-Clarity™ quality level = MQ100

P10402-25G P10402-5G
SIALP10402-25GEA 9410 CZK
SIALP10402-25G SIALP10402-5G
1-Iodoperfluoropropane, Sigma-Aldrich®
1-Iodoperfluoropropane
Vzorec: C₃F₇I
MW: 295,93 g/mol
Bod varu: 41 °C (1013 hPa)
Bod tání: –95 °C
Hustota: 2,1306 g/cm³ (0 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001061
CAS číslo: 754-34-7
EINECS: 212-045-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR