Tisk…

Gadolinium trinitrate hexahydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Gadolinium (III) nitrate hexahydrate

M-Clarity™ quality level = MQ200

451134-10G 451134-50G
SIAL451134-10GEA 4920 CZK
SIAL451134-10G SIAL451134-50G
Gadolinium trinitrate hexahydrate, Sigma-Aldrich®
Gadolinium trinitrate hexahydrate
Vzorec: GdN₃O₉·6H₂O
Hustota: 2,4 g/cm³ (20 °C)
MDL Number: MFCD00149728
CAS číslo: 19598-90-4
EINECS: 233-437-2
UN: 1477
ADR: 5.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR