Gadolinium trinitrate pentahydrate ≥99.9%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
GADOLINIUM NITRATE, Gadolinium(III) nitrate hydrate
12917.14
12917.22
12917.14EA
2180
CZK
12917.14
12917.22
Gadolinium trinitrate pentahydrate ≥99.9%
Gadolinium(III) nitrate hydrate
Purity: 99.9%
CAS No.: 94219-55-3
Molecular Formula: Gd(NO3)3.xH2O (x≈6)
Formula Weight: 343.26(anhy)
Physical Form: Crystalline
Density: 2.406
Appearance: White
Melting Point: 92°
Merck Reference: 14,4326
UN No.: 1477
MDL No.: MFCD00011023
Vzorec:
Gd(NO₃)₃·xH₂O MW: 343.26 g/mol Hustota: 2.406 Storage Temperature: Ambient |
MDL Number:
MFCD00149730 CAS číslo: 94219-55-3 EINECS: 233-437-2 UN: 1477 ADR: 5.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...